Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTGGGAAGAGGGGAGGTGGCGAG[A/G]GCTGAGGAAGCGGGTAGGGAAAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608550 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCED1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PCED1A - PC-esterase domain containing 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271168.1 | 1285 | Missense Mutation | CCC,CTC | P,L 280 | NP_001258097.1 | |
NM_022760.4 | 1285 | Missense Mutation | CCC,CTC | P,L 331 | NP_073597.2 | |
XM_005260804.2 | 1285 | Missense Mutation | CCC,CTC | P,L 331 | XP_005260861.1 | |
XM_005260805.2 | 1285 | Missense Mutation | CCC,CTC | P,L 280 | XP_005260862.1 | |
XM_005260806.3 | 1285 | Intron | XP_005260863.1 |
VPS16 - VPS16, CORVET/HOPS core subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |