Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTAGCCGCAATCCTCGCAGACGT[A/G]GAGCTTGTCCCGCCGCTGCTTATAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616441 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OVOL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OVOL2 - ovo like zinc finger 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303461.1 | 888 | Missense Mutation | CAC,TAC | H,Y 82 | NP_001290390.1 | |
NM_001303462.1 | 888 | Missense Mutation | CAC,TAC | H,Y 82 | NP_001290391.1 | |
NM_021220.3 | 888 | Missense Mutation | CAC,TAC | H,Y 214 | NP_067043.2 |