Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAGATGAGGGAACAAAGCGGCTA[A/G]CAGTGGGACACAAACAGCTCTTAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614124 | ||||||||||||||||||||
Literature Links: |
GPCPD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPCPD1 - glycerophosphocholine phosphodiesterase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019593.3 | 2298 | Missense Mutation | GCT,GTT | A,V 645 | NP_062539.1 | |
XM_005260758.4 | 2298 | Missense Mutation | GCT,GTT | A,V 645 | XP_005260815.1 | |
XM_005260761.1 | 2298 | Missense Mutation | GCT,GTT | A,V 464 | XP_005260818.1 | |
XM_006723596.3 | 2298 | Missense Mutation | GCT,GTT | A,V 645 | XP_006723659.1 |