Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCAAATGCGTAGTTTCCAGTTTC[A/T]TGACGGCAAAGAGCACAGGTATGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614289 | ||||||||||||||||||||
Literature Links: |
SEL1L2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SEL1L2 - SEL1L2 ERAD E3 ligase adaptor subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271539.1 | 1739 | Missense Mutation | AAG,ATG | K,M 521 | NP_001258468.1 | |
XM_006723646.2 | 1739 | Missense Mutation | AAG,ATG | K,M 634 | XP_006723709.1 | |
XM_006723647.2 | 1739 | Missense Mutation | AAG,ATG | K,M 634 | XP_006723710.1 | |
XM_006723648.2 | 1739 | Missense Mutation | AAG,ATG | K,M 615 | XP_006723711.1 | |
XM_006723650.2 | 1739 | Silent Mutation | CAA,CAT | Q,H 576 | XP_006723713.1 | |
XM_006723651.2 | 1739 | Silent Mutation | CAA,CAT | Q,H 576 | XP_006723714.1 | |
XM_006723652.2 | 1739 | Missense Mutation | AAG,ATG | K,M 521 | XP_006723715.1 | |
XM_006723654.3 | 1739 | Missense Mutation | AAG,ATG | K,M 298 | XP_006723717.1 | |
XM_006723655.2 | 1739 | Intron | XP_006723718.1 | |||
XM_006723656.2 | 1739 | Intron | XP_006723719.1 | |||
XM_011529374.1 | 1739 | Missense Mutation | AAG,ATG | K,M 614 | XP_011527676.1 | |
XM_011529375.1 | 1739 | Missense Mutation | AAG,ATG | K,M 597 | XP_011527677.1 | |
XM_011529376.1 | 1739 | Missense Mutation | AAG,ATG | K,M 578 | XP_011527678.1 | |
XM_011529377.1 | 1739 | Missense Mutation | AAG,ATG | K,M 549 | XP_011527679.1 | |
XM_011529378.1 | 1739 | Missense Mutation | AAG,ATG | K,M 522 | XP_011527680.1 | |
XM_011529379.1 | 1739 | Missense Mutation | AAG,ATG | K,M 495 | XP_011527681.1 | |
XM_017028084.1 | 1739 | Missense Mutation | AAG,ATG | K,M 283 | XP_016883573.1 |