Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTGGCTGGGGCCCCTCCAGTGCC[A/G]TGGAGTTGGAGCCTCCTTGAGCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605020 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
VSX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VSX1 - visual system homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256271.1 | 1338 | Intron | NP_001243200.1 | |||
NM_001256272.1 | 1338 | Intron | NP_001243201.1 | |||
NM_014588.5 | 1338 | Missense Mutation | NP_055403.2 | |||
NM_199425.2 | 1338 | Intron | NP_955457.1 | |||
XM_017027837.1 | 1338 | Intron | XP_016883326.1 | |||
XM_017027838.1 | 1338 | Intron | XP_016883327.1 |