Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTTGCTATAATGGCTTCTTTTAC[A/G]TCTGATTTCTGGCAGGAGGTCCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600490 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NFATC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NFATC2 - nuclear factor of activated T-cells 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136021.2 | 2295 | UTR 3 | NP_001129493.1 | |||
NM_001258292.1 | 2295 | Missense Mutation | ACG,ATG | T,M 905 | NP_001245221.1 | |
NM_001258294.1 | 2295 | UTR 3 | NP_001245223.1 | |||
NM_001258295.1 | 2295 | Missense Mutation | ACG,ATG | T,M 706 | NP_001245224.1 | |
NM_001258296.1 | 2295 | UTR 3 | NP_001245225.1 | |||
NM_001258297.1 | 2295 | Missense Mutation | ACG,ATG | T,M 706 | NP_001245226.1 | |
NM_012340.4 | 2295 | UTR 3 | NP_036472.2 | |||
NM_173091.3 | 2295 | Missense Mutation | ACG,ATG | T,M 925 | NP_775114.1 | |
XM_011528824.2 | 2295 | Intron | XP_011527126.1 | |||
XM_011528825.2 | 2295 | Intron | XP_011527127.1 | |||
XM_011528826.2 | 2295 | Intron | XP_011527128.1 | |||
XM_017027850.1 | 2295 | Intron | XP_016883339.1 | |||
XM_017027851.1 | 2295 | Missense Mutation | ACG,ATG | T,M 695 | XP_016883340.1 |