Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCTCCAGCATGCCCATCTGCACC[A/G]TCTCCATGAGCCAGGACTTCGGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611625 MIM: 612107 | ||||||||||||||||||||
Literature Links: |
GID8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GID8 - GID complex subunit 8 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC17A9 - solute carrier family 17 member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302643.1 | 391 | Missense Mutation | ATC,GTC | I,V 43 | NP_001289572.1 | |
NM_022082.3 | 391 | Missense Mutation | ATC,GTC | I,V 49 | NP_071365.3 | |
XM_011528978.2 | 391 | UTR 5 | XP_011527280.1 |