Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGGGCTACACCTTGCAGATCTTC[A/G]AGCTCACTGGCTCTGGGCGCTTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
COL20A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COL20A1 - collagen type XX alpha 1 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020882.2 | 406 | Missense Mutation | AAG,GAG | K,E 97 | NP_065933.2 | |
XM_011528937.1 | 406 | Missense Mutation | AAG,GAG | K,E 97 | XP_011527239.1 | |
XM_011528938.1 | 406 | Missense Mutation | AAG,GAG | K,E 97 | XP_011527240.1 | |
XM_011528939.1 | 406 | Missense Mutation | AAG,GAG | K,E 97 | XP_011527241.1 | |
XM_011528940.1 | 406 | Missense Mutation | AAG,GAG | K,E 97 | XP_011527242.1 |