Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCTCCCTTGCCTGGTCATGGTTT[C/T]CTGGTGAGAAGAGTGTTCCTGGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610302 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EDEM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EDEM2 - ER degradation enhancing alpha-mannosidase like protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145025.1 | 1602 | Missense Mutation | GAA,GGA | E,G 501 | NP_001138497.1 | |
NM_018217.2 | 1602 | Missense Mutation | GAA,GGA | E,G 538 | NP_060687.2 |