Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCAGTTTCAGCCAGTTCCATCTC[A/G]TTGTGAACTGCTGGCAAGAGACATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614289 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEL1L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEL1L2 - SEL1L2 ERAD E3 ligase adaptor subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271539.1 | 1796 | Missense Mutation | ACG,ATG | T,M 539 | NP_001258468.1 | |
XM_006723646.2 | 1796 | Missense Mutation | ACG,ATG | T,M 653 | XP_006723709.1 | |
XM_006723647.2 | 1796 | Missense Mutation | ACG,ATG | T,M 652 | XP_006723710.1 | |
XM_006723648.2 | 1796 | Missense Mutation | ACG,ATG | T,M 634 | XP_006723711.1 | |
XM_006723650.2 | 1796 | UTR 3 | XP_006723713.1 | |||
XM_006723651.2 | 1796 | UTR 3 | XP_006723714.1 | |||
XM_006723652.2 | 1796 | Missense Mutation | ACG,ATG | T,M 540 | XP_006723715.1 | |
XM_006723654.3 | 1796 | Missense Mutation | ACG,ATG | T,M 317 | XP_006723717.1 | |
XM_006723655.2 | 1796 | Intron | XP_006723718.1 | |||
XM_006723656.2 | 1796 | Intron | XP_006723719.1 | |||
XM_011529374.1 | 1796 | Missense Mutation | ACG,ATG | T,M 633 | XP_011527676.1 | |
XM_011529375.1 | 1796 | Missense Mutation | ACG,ATG | T,M 616 | XP_011527677.1 | |
XM_011529376.1 | 1796 | Missense Mutation | ACG,ATG | T,M 597 | XP_011527678.1 | |
XM_011529377.1 | 1796 | Missense Mutation | ACG,ATG | T,M 568 | XP_011527679.1 | |
XM_011529378.1 | 1796 | Missense Mutation | ACG,ATG | T,M 541 | XP_011527680.1 | |
XM_011529379.1 | 1796 | Missense Mutation | ACG,ATG | T,M 514 | XP_011527681.1 | |
XM_017028084.1 | 1796 | Missense Mutation | ACG,ATG | T,M 302 | XP_016883573.1 |