Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGAGGCTGGGCTGCCGGGACTGC[C/T]GGGTGTGGATGTGAGTGCGCCTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 120270 MIM: 606459 | ||||||||||||||||||||
Literature Links: |
COL9A3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COL9A3 - collagen type IX alpha 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001853.3 | 246 | Missense Mutation | CCG,CTG | P,L 82 | NP_001844.3 | |
XM_011528545.1 | 246 | Missense Mutation | CCG,CTG | P,L 82 | XP_011526847.1 | |
XM_017027666.1 | 246 | Missense Mutation | CCG,CTG | P,L 82 | XP_016883155.1 |
OGFR - opioid growth factor receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |