Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCAAGCTATGAGGACAGCAGCCT[A/G]CCCTCCATCAATGCGCTCCTGCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600281 | ||||||||||||||||||||
Literature Links: |
HNF4A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HNF4A - hepatocyte nuclear factor 4 alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000457.4 | 607 | Silent Mutation | CTA,CTG | L,L 149 | NP_000448.3 | |
NM_001030003.2 | 607 | Silent Mutation | CTA,CTG | L,L 127 | NP_001025174.1 | |
NM_001030004.2 | 607 | Silent Mutation | CTA,CTG | L,L 127 | NP_001025175.1 | |
NM_001258355.1 | 607 | Silent Mutation | CTA,CTG | L,L 142 | NP_001245284.1 | |
NM_001287182.1 | 607 | Silent Mutation | CTA,CTG | L,L 124 | NP_001274111.1 | |
NM_001287183.1 | 607 | Silent Mutation | CTA,CTG | L,L 124 | NP_001274112.1 | |
NM_001287184.1 | 607 | Silent Mutation | CTA,CTG | L,L 124 | NP_001274113.1 | |
NM_175914.4 | 607 | Silent Mutation | CTA,CTG | L,L 127 | NP_787110.2 | |
NM_178849.2 | 607 | Silent Mutation | CTA,CTG | L,L 149 | NP_849180.1 | |
NM_178850.2 | 607 | Silent Mutation | CTA,CTG | L,L 149 | NP_849181.1 | |
XM_005260407.3 | 607 | Silent Mutation | CTA,CTG | L,L 188 | XP_005260464.1 |
MIR3646 - microRNA 3646 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |