Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTTGGTCAGGACATCTTTTGTTC[G/T]ATCTTCAGGCCGCAGAAGTCCCCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610924 MIM: 611663 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBCK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RBCK1 - RANBP2-type and C3HC4-type zinc finger containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBC1D20 - TBC1 domain family member 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144628.3 | 979 | Silent Mutation | NP_653229.1 | |||
XM_005260661.1 | 979 | Missense Mutation | XP_005260718.1 | |||
XM_006723540.3 | 979 | Missense Mutation | XP_006723603.1 | |||
XM_017027645.1 | 979 | Missense Mutation | XP_016883134.1 |