Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTGCAGCGAGCCTTAAGGTTCTT[G/T]CTGGGTCGCTTGCCCTGGCGAGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601146 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GDF5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GDF5 - growth differentiation factor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000557.4 | 1441 | Missense Mutation | AGA,AGC | R,S 393 | NP_000548.2 | |
NM_001319138.1 | 1441 | Missense Mutation | AGA,AGC | R,S 393 | NP_001306067.1 | |
XM_011529075.2 | 1441 | Missense Mutation | AGA,AGC | R,S 393 | XP_011527377.1 |