Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGACCGTGGGGCGCTTCAGGGT[C/G]CGGCTTTTCTGAGGTTCCAAGCATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602523 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DSCAM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DSCAM - DS cell adhesion molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271534.1 | 6257 | Silent Mutation | CGC,CGG | R,R 1917 | NP_001258463.1 | |
NM_001389.3 | 6257 | Silent Mutation | CGC,CGG | R,R 1935 | NP_001380.2 | |
XM_017028281.1 | 6257 | Silent Mutation | CGC,CGG | R,R 1699 | XP_016883770.1 |