Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTCACGGGCACACACACGGCTGG[C/G]TTGAAGCTCACGGGCACGCACACGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612920 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KRTAP12-1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KRTAP12-1 - keratin associated protein 12-1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181686.1 | 163 | Missense Mutation | AAC,AAG | N,K 41 | NP_859014.1 |
TSPEAR - thrombospondin type laminin G domain and EAR repeats | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272037.1 | 163 | Intron | NP_001258966.1 | |||
NM_144991.2 | 163 | Intron | NP_659428.2 |