Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCACCGCAATGGCGGCTGTGAG[A/G]GTCCTGGTGGCCTCGAGGCTCGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601659 MIM: 601475 | ||||||||||||||||||||
Literature Links: |
C21orf33 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C21orf33 - chromosome 21 open reading frame 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320383.1 | Intron | NP_001307312.1 | ||||
NM_001320384.1 | Intron | NP_001307313.1 | ||||
NM_004649.7 | Intron | NP_004640.4 | ||||
NM_198155.4 | Intron | NP_937798.4 | ||||
XM_017028479.1 | Intron | XP_016883968.1 |
PWP2 - PWP2 periodic tryptophan protein homolog (yeast) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |