Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGCGACCTGAGCTACAGCAGCC[A/G]CGTCTGCCTTCCTGGTTCCTGTGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612920 | ||||||||||||||||||||
Literature Links: |
KRTAP10-4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KRTAP10-4 - keratin associated protein 10-4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198687.2 | 68 | Missense Mutation | CAC,CGC | H,R 13 | NP_941960.2 |
KRTAP10-5 - keratin associated protein 10-5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSPEAR - thrombospondin type laminin G domain and EAR repeats | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272037.1 | 68 | Intron | NP_001258966.1 | |||
NM_144991.2 | 68 | Intron | NP_659428.2 |