Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTCAACACTGACCCATTTATTT[C/G]CATGCTTTCAATCAGATTCTTGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138440 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GART PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GART - phosphoribosylglycinamide formyltransferase, phosphoribosylglycinamide synthetase, phosphoribosylaminoimidazole synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000819.4 | 2667 | Missense Mutation | CAA,GAA | Q,E 786 | NP_000810.1 | |
NM_001136005.1 | 2667 | Missense Mutation | CAA,GAA | Q,E 786 | NP_001129477.1 | |
NM_001136006.1 | 2667 | Missense Mutation | CAA,GAA | Q,E 786 | NP_001129478.1 | |
NM_175085.2 | 2667 | Intron | NP_780294.1 | |||
XM_005260941.1 | 2667 | Missense Mutation | CAA,GAA | Q,E 786 | XP_005260998.1 | |
XM_006723989.1 | 2667 | Missense Mutation | CAA,GAA | Q,E 786 | XP_006724052.1 | |
XM_006723990.1 | 2667 | Missense Mutation | CAA,GAA | Q,E 786 | XP_006724053.1 | |
XM_011529526.1 | 2667 | Missense Mutation | CAA,GAA | Q,E 786 | XP_011527828.1 |