Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCTGCTGCAGGAAGGTCGTGTA[C/T]TATATTCGGTTGTTCTAGCAATGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605296 | ||||||||||||||||||||
Literature Links: |
BRWD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BRWD1 - bromodomain and WD repeat domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSMG1 - proteasome assembly chaperone 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261824.1 | 700 | Missense Mutation | ATA,GTA | I,V 207 | NP_001248753.1 | |
NM_001320795.1 | 700 | Missense Mutation | ATA,GTA | I,V 125 | NP_001307724.1 | |
NM_003720.3 | 700 | Missense Mutation | ATA,GTA | I,V 212 | NP_003711.1 | |
NM_203433.2 | 700 | Missense Mutation | ATA,GTA | I,V 191 | NP_982257.1 | |
XM_017028488.1 | 700 | Missense Mutation | ATA,GTA | I,V 104 | XP_016883977.1 |