Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGCGACTTCCTGCGCAAGGAGTAC[A/G]GGGGCCTGGACGTGCTGGTCAACAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 114830 | ||||||||||||||||||||
Literature Links: |
CBR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CBR1 - carbonyl reductase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286789.1 | 436 | Missense Mutation | AGG,GGG | R,G 82 | NP_001273718.1 | |
NM_001757.3 | 436 | Missense Mutation | AGG,GGG | R,G 82 | NP_001748.1 |
LOC100133286 - uncharacterized LOC100133286 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SETD4 - SET domain containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |