Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCACTTGTCTTTCTCGGTGCCGAA[A/G]ATGGAGGCCAGATACTCCGCCATTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 191317 | ||||||||||||||||||||
Literature Links: |
U2AF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
U2AF1 - U2 small nuclear RNA auxiliary factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025203.1 | Intron | NP_001020374.1 | ||||
NM_001025204.1 | Intron | NP_001020375.1 | ||||
NM_006758.2 | Intron | NP_006749.1 | ||||
XM_011529743.2 | Intron | XP_011528045.1 | ||||
XM_017028468.1 | Intron | XP_016883957.1 |