Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAGAGGCCCCCCGGCCAGCCTTC[A/G]TGGGCTCAGCCGTGTCTGAGTTGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 165095 | ||||||||||||||||||||
Literature Links: |
OSM PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OSM - oncostatin M | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319108.1 | 541 | Missense Mutation | ACG,ATG | T,M 145 | NP_001306037.1 | |
NM_020530.5 | 541 | Missense Mutation | ACG,ATG | T,M 166 | NP_065391.1 |