Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600574 MIM: 608077 MIM: 609518 | ||||||||||||||||||||
Literature Links: |
LZTR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LZTR1 - leucine zipper like transcription regulator 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
P2RX6 - purinergic receptor P2X 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
THAP7 - THAP domain containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008695.1 | 379 | Missense Mutation | CAG,GAG | Q,E 59 | NP_001008695.1 | |
NM_030573.2 | 379 | Missense Mutation | CAG,GAG | Q,E 59 | NP_085050.2 |
THAP7-AS1 - THAP7 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TUBA3FP - tubulin alpha 3f pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |