Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGACGGACTGAGCCAGAGGCCCTG[A/G]AGCCAGGGGGGAACCTAGTGGCTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613777 MIM: 609063 | ||||||||||||||||||||
Literature Links: |
FOXRED2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FOXRED2 - FAD dependent oxidoreductase domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102371.1 | 1871 | Missense Mutation | CCA,TCA | P,S 670 | NP_001095841.1 | |
NM_024955.5 | 1871 | Missense Mutation | CCA,TCA | P,S 670 | NP_079231.4 | |
XM_017028941.1 | 1871 | Missense Mutation | CCA,TCA | P,S 580 | XP_016884430.1 | |
XM_017028942.1 | 1871 | Missense Mutation | CCA,TCA | P,S 580 | XP_016884431.1 |
TXN2 - thioredoxin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |