Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTGGGCCAGCAGTGCCCTGGAC[A/G]TGGCCTCGAACCGCGTGCCCTGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607922 | ||||||||||||||||||||
Literature Links: |
A4GALT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
A4GALT - alpha 1,4-galactosyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318038.1 | 1474 | Missense Mutation | ACG,ATG | T,M 329 | NP_001304967.1 | |
NM_017436.5 | 1474 | Missense Mutation | ACG,ATG | T,M 329 | NP_059132.1 | |
XM_005261644.2 | 1474 | Missense Mutation | ACG,ATG | T,M 329 | XP_005261701.1 | |
XM_005261646.4 | 1474 | Missense Mutation | ACG,ATG | T,M 329 | XP_005261703.1 | |
XM_005261647.2 | 1474 | Missense Mutation | ACG,ATG | T,M 329 | XP_005261704.1 | |
XM_005261648.3 | 1474 | Missense Mutation | ACG,ATG | T,M 329 | XP_005261705.1 | |
XM_006724265.3 | 1474 | Missense Mutation | ACG,ATG | T,M 329 | XP_006724328.1 | |
XM_006724266.3 | 1474 | Missense Mutation | ACG,ATG | T,M 329 | XP_006724329.1 | |
XM_011530233.2 | 1474 | Missense Mutation | ACG,ATG | T,M 329 | XP_011528535.1 | |
XM_017028831.1 | 1474 | Missense Mutation | ACG,ATG | T,M 329 | XP_016884320.1 |