Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGTGTCCAGGGAGGGGGCCCCTC[A/G]GCCAGGTCGGCATCTGCTGCAGCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608457 | ||||||||||||||||||||
Literature Links: |
CBX7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CBX7 - chromobox 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175709.3 | 648 | Silent Mutation | GCC,GCT | A,A 205 | NP_783640.1 | |
XM_005261413.4 | 648 | Silent Mutation | GCC,GCT | A,A 112 | XP_005261470.1 | |
XM_006724174.3 | 648 | Silent Mutation | GCC,GCT | A,A 213 | XP_006724237.1 | |
XM_006724175.3 | 648 | Silent Mutation | GCC,GCT | A,A 212 | XP_006724238.1 | |
XM_006724176.3 | 648 | Silent Mutation | GCC,GCT | A,A 154 | XP_006724239.1 | |
XM_006724177.3 | 648 | Silent Mutation | GCC,GCT | A,A 146 | XP_006724240.1 | |
XM_006724178.3 | 648 | Silent Mutation | GCC,GCT | A,A 120 | XP_006724241.1 | |
XM_011530025.2 | 648 | Silent Mutation | GCC,GCT | A,A 119 | XP_011528327.1 | |
XM_017028679.1 | 648 | Silent Mutation | GCC,GCT | A,A 204 | XP_016884168.1 |