Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGGGGGCATCTAGGGGCAGGCGG[A/G]TGGATGGTGAAGGTTGTAGGGCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 617034 MIM: 610020 | ||||||||||||||||||||
Literature Links: |
GATSL3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GATSL3 - GATS protein like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBC1D10A - TBC1 domain family member 10A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204240.1 | 1335 | Missense Mutation | ACC,ATC | T,I 417 | NP_001191169.1 | |
NM_031937.2 | 1335 | Missense Mutation | ACC,ATC | T,I 410 | NP_114143.1 |