Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATGAGGGAAAGGGGCTCTGGCCC[A/G]TCATGCCCACTGGTCAGCAGGGGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606078 MIM: 610440 | ||||||||||||||||||||
Literature Links: |
MKL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MKL1 - megakaryoblastic leukemia (translocation) 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282660.1 | 3016 | Silent Mutation | GAC,GAT | D,D 832 | NP_001269589.1 | |
NM_001282661.1 | 3016 | Silent Mutation | GAC,GAT | D,D 782 | NP_001269590.1 | |
NM_001282662.1 | 3016 | UTR 3 | NP_001269591.1 | |||
NM_001318139.1 | 3016 | Silent Mutation | GAC,GAT | D,D 867 | NP_001305068.1 | |
NM_020831.4 | 3016 | Silent Mutation | GAC,GAT | D,D 832 | NP_065882.1 | |
XM_005261692.1 | 3016 | Silent Mutation | GAC,GAT | D,D 859 | XP_005261749.1 | |
XM_005261694.1 | 3016 | Silent Mutation | GAC,GAT | D,D 832 | XP_005261751.1 | |
XM_011530283.1 | 3016 | Silent Mutation | GAC,GAT | D,D 859 | XP_011528585.1 | |
XM_011530285.1 | 3016 | Silent Mutation | GAC,GAT | D,D 791 | XP_011528587.1 | |
XM_011530287.1 | 3016 | Silent Mutation | GAC,GAT | D,D 667 | XP_011528589.1 | |
XM_017028888.1 | 3016 | Silent Mutation | GAC,GAT | D,D 859 | XP_016884377.1 | |
XM_017028889.1 | 3016 | Silent Mutation | GAC,GAT | D,D 791 | XP_016884378.1 |
SGSM3 - small G protein signaling modulator 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |