Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGCCTGTGAACTATGGGAACAGA[C/T]TCTCCAGGACCACCCGACAGACATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TTC38 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TTC38 - tetratricopeptide repeat domain 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017931.2 | 477 | Missense Mutation | ACT,ATT | T,I 134 | NP_060401.2 | |
XM_011530259.1 | 477 | Missense Mutation | ACT,ATT | T,I 69 | XP_011528561.1 | |
XM_011530260.2 | 477 | Missense Mutation | ACT,ATT | T,I 134 | XP_011528562.1 |