Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTCCGCAAGTGGACCACCACCGC[A/G]CTTGGGCTGTTGACCATCCCCATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610235 MIM: 607558 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTFP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MTFP1 - mitochondrial fission process 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003704.2 | 641 | Intron | NP_001003704.1 | |||
NM_016498.4 | 641 | Silent Mutation | GCA,GCG | A,A 128 | NP_057582.2 |
SEC14L2 - SEC14 like lipid binding 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |