Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACGGCGCTCACGGTGAGCGCCCG[C/T]GCCGCCTGCACCTCGGTGCTCAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603465 MIM: 602101 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDC45 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDC45 - cell division cycle 45 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CLDN5 - claudin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130861.1 | 710 | Silent Mutation | GCA,GCG | A,A 165 | NP_001124333.1 | |
NM_003277.3 | 710 | Silent Mutation | GCA,GCG | A,A 165 | NP_003268.2 | |
XM_017028929.1 | 710 | Silent Mutation | GCA,GCG | A,A 165 | XP_016884418.1 | |
XM_017028930.1 | 710 | Silent Mutation | GCA,GCG | A,A 165 | XP_016884419.1 |