Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAACATGTCAGGGTGGGAGTCAT[A/G]TTACAAAACCGAGGGCGATGAAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614637 MIM: 152690 | ||||||||||||||||||||
Literature Links: |
DESI1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DESI1 - desumoylating isopeptidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015704.2 | Intron | NP_056519.1 | ||||
XM_005261571.4 | Intron | XP_005261628.1 |
XRCC6 - X-ray repair cross complementing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288976.1 | Intron | NP_001275905.1 | ||||
NM_001288977.1 | Intron | NP_001275906.1 | ||||
NM_001288978.1 | Intron | NP_001275907.1 | ||||
NM_001469.4 | Intron | NP_001460.1 |