Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTATGTCCTGGGCACTGCCACCC[A/G]CTGGCCCCTGGCTGTCCGCAAGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610235 MIM: 607558 | ||||||||||||||||||||
Literature Links: |
MTFP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MTFP1 - mitochondrial fission process 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003704.2 | 604 | Intron | NP_001003704.1 | |||
NM_016498.4 | 604 | Missense Mutation | CAC,CGC | H,R 116 | NP_057582.2 |
SEC14L2 - SEC14 like lipid binding 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |