Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGATCCGCAGCTCCGCCAGCACCC[A/G]GATGCCGTTGGTCAGCTTCAGGGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600157 | ||||||||||||||||||||
Literature Links: |
AP1B1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AP1B1 - adaptor related protein complex 1 beta 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127.3 | 2885 | Missense Mutation | CGG,TGG | R,W 907 | NP_001118.3 | |
NM_001166019.1 | 2885 | Missense Mutation | CGG,TGG | R,W 880 | NP_001159491.1 | |
NM_145730.2 | 2885 | Missense Mutation | CGG,TGG | R,W 900 | NP_663782.2 |
MIR3653 - microRNA 3653 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD125 - small nucleolar RNA, C/D box 125 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |