Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGCTCTTCAGTTTCAGAGATGCA[A/G]TGAAAAATGCCAGAGTCACTACACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611674 | ||||||||||||||||||||
Literature Links: |
XKR3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
XKR3 - XK related 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318251.1 | 873 | Missense Mutation | ACT,ATT | T,I 257 | NP_001305180.1 | |
NM_175878.4 | 873 | Missense Mutation | ACT,ATT | T,I 257 | NP_787074.2 | |
XM_017028600.1 | 873 | Missense Mutation | ACT,ATT | T,I 257 | XP_016884089.1 |