Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACTAAGCCAGACCATGCTGGGCC[A/G]CCCCAACTCACACAGCAACTCCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606021 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRAME PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRAME - preferentially expressed antigen in melanoma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291715.1 | 1951 | Silent Mutation | NP_001278644.1 | |||
NM_001291716.1 | 1951 | Silent Mutation | NP_001278645.1 | |||
NM_001291717.1 | 1951 | Silent Mutation | NP_001278646.1 | |||
NM_001291719.1 | 1951 | Silent Mutation | NP_001278648.1 | |||
NM_001318126.1 | 1951 | Silent Mutation | NP_001305055.1 | |||
NM_001318127.1 | 1951 | Silent Mutation | NP_001305056.1 | |||
NM_006115.4 | 1951 | Silent Mutation | NP_006106.1 | |||
NM_206953.2 | 1951 | Silent Mutation | NP_996836.1 | |||
NM_206954.2 | 1951 | Silent Mutation | NP_996837.1 | |||
NM_206955.2 | 1951 | Silent Mutation | NP_996838.1 | |||
NM_206956.2 | 1951 | Silent Mutation | NP_996839.1 | |||
XM_011530034.2 | 1951 | Missense Mutation | XP_011528336.1 |