Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAGCTGTCTTCCGGAAGAGGGGC[C/G]TCTTCCGAAACCAGGTAGCACCAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609900 MIM: 608993 | ||||||||||||||||||||
Literature Links: |
APOBEC3D PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
APOBEC3D - apolipoprotein B mRNA editing enzyme catalytic subunit 3D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152426.3 | Intron | NP_689639.2 | ||||
XM_005261337.3 | Intron | XP_005261394.1 | ||||
XM_017028596.1 | Intron | XP_016884085.1 |
APOBEC3F - apolipoprotein B mRNA editing enzyme catalytic subunit 3F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006666.1 | Intron | NP_001006667.1 | ||||
NM_145298.5 | Intron | NP_660341.2 | ||||
XM_011529994.2 | Intron | XP_011528296.1 | ||||
XM_017028642.1 | Intron | XP_016884131.1 | ||||
XM_017028643.1 | Intron | XP_016884132.1 |