Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGTGAATCAGAGCCACACCGAGAA[C/G]CGCCGCGGAGCCCTCATCCCTAACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608314 MIM: 610981 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEPT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEPT3 - septin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019106.5 | 124 | Intron | NP_061979.3 | |||
NM_145733.2 | 124 | Intron | NP_663786.2 | |||
XM_017028862.1 | 124 | Intron | XP_016884351.1 |
WBP2NL - WBP2 N-terminal like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152613.2 | 124 | Missense Mutation | AAC,AAG | N,K 10 | NP_689826.2 |