Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGAAGCGAGATGGTGACGACCGG[C/T]GGCCGAGGCTCGTTCTTAGCTTCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 150570 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LGALS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LGALS1 - galectin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NOL12 - nucleolar protein 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024313.2 | 99 | Missense Mutation | CGG,TGG | R,W 15 | NP_077289.1 |