Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGCTGGCCATTGCTGTGCTGGCC[A/G]TCCTGGGCAATGTGCTGGTGTGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102776 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADORA2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADORA2A - adenosine A2a receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000675.5 | 691 | Missense Mutation | ATC,GTC | I,V 21 | NP_000666.2 | |
NM_001278497.1 | 691 | Missense Mutation | ATC,GTC | I,V 21 | NP_001265426.1 | |
NM_001278498.1 | 691 | Missense Mutation | ATC,GTC | I,V 21 | NP_001265427.1 | |
NM_001278499.1 | 691 | Missense Mutation | ATC,GTC | I,V 21 | NP_001265428.1 | |
NM_001278500.1 | 691 | Missense Mutation | ATC,GTC | I,V 21 | NP_001265429.1 |
ADORA2A-AS1 - ADORA2A antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPECC1L-ADORA2A - SPECC1L-ADORA2A readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |