Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCATTCGCAATGCAGCCGATATCT[C/T]CATCATCGTTATCTACTTCGTGGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 182380 | ||||||||||||||||||||
Literature Links: |
SLC5A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC5A1 - solute carrier family 5 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000343.3 | 339 | Missense Mutation | TCC,TTC | S,F 30 | NP_000334.1 | |
NM_001256314.1 | 339 | Intron | NP_001243243.1 | |||
XM_011530331.1 | 339 | Missense Mutation | TCC,TTC | S,F 30 | XP_011528633.1 |