Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTACTCCCTGCTGTTCCGCAGGA[C/T]CTCCACCTTCGCCCTCACCATCATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610843 | ||||||||||||||||||||
Literature Links: |
UQCR10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UQCR10 - ubiquinol-cytochrome c reductase, complex III subunit X | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003684.1 | 83 | Missense Mutation | ACC,ATC | T,I 18 | NP_001003684.1 | |
NM_013387.3 | 83 | Missense Mutation | ACC,ATC | T,I 18 | NP_037519.2 |
ZMAT5 - zinc finger matrin-type 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003692.1 | 83 | Intron | NP_001003692.1 | |||
NM_001318129.1 | 83 | Intron | NP_001305058.1 | |||
NM_019103.2 | 83 | Intron | NP_061976.1 |