Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACCGATCTCACTCCCTGCTTGAA[C/T]TGCGCCATGTCGGGGGCGTTGGGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612439 | ||||||||||||||||||||
Literature Links: |
ARFGAP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARFGAP3 - ADP ribosylation factor GTPase activating protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142293.1 | 1681 | Silent Mutation | CAA,CAG | Q,Q 443 | NP_001135765.1 | |
NM_014570.4 | 1681 | Silent Mutation | CAA,CAG | Q,Q 487 | NP_055385.3 | |
XM_005261525.3 | 1681 | Silent Mutation | CAA,CAG | Q,Q 443 | XP_005261582.1 |