Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCGGGGACGCTGTCAACAACACC[A/G]AGTGGTGAGTGAGGCCTGCTGAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138720 MIM: 602724 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GP1BB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GP1BB - glycoprotein Ib platelet beta subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEPT5 - septin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009939.2 | 688 | Missense Mutation | AAG,GAG | K,E 129 | NP_001009939.1 | |
NM_002688.5 | 688 | Missense Mutation | AAG,GAG | K,E 120 | NP_002679.2 |
SEPT5-GP1BB - SEPT5-GP1BB readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |