Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACCTCGGTGCTCAGAGCCAGCAC[C/G]GAGTCGTACACTTTGCACTGCATGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603465 MIM: 602101 | ||||||||||||||||||||
Literature Links: |
CDC45 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDC45 - cell division cycle 45 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CLDN5 - claudin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130861.1 | 677 | Silent Mutation | TCC,TCG | S,S 154 | NP_001124333.1 | |
NM_003277.3 | 677 | Silent Mutation | TCC,TCG | S,S 154 | NP_003268.2 | |
XM_017028929.1 | 677 | Silent Mutation | TCC,TCG | S,S 154 | XP_016884418.1 | |
XM_017028930.1 | 677 | Silent Mutation | TCC,TCG | S,S 154 | XP_016884419.1 |