Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCACCTTCTCATCTCCACAGTCTC[A/G]CCTGGAAGACAAGGGACACACAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603107 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCF20 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TCF20 - transcription factor 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005650.3 | 6250 | UTR 3 | NP_005641.1 | |||
NM_181492.2 | 6250 | Missense Mutation | GCG,GTG | A,V 1934 | NP_852469.1 | |
XM_005261722.3 | 6250 | UTR 3 | XP_005261779.1 | |||
XM_006724313.3 | 6250 | UTR 3 | XP_006724376.1 | |||
XM_011530353.2 | 6250 | UTR 3 | XP_011528655.1 | |||
XM_011530354.2 | 6250 | Missense Mutation | GCG,GTG | A,V 1934 | XP_011528656.1 |