Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCGTAGCGCACCATGGCGTTCTT[C/G]TCCTGCACTAGCTTGAACCACTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608882 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MICAL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MICAL3 - microtubule associated monooxygenase, calponin and LIM domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122731.2 | 6428 | Intron | NP_001116203.1 | |||
NM_001136004.3 | 6428 | Intron | NP_001129476.1 | |||
NM_015241.2 | 6428 | Missense Mutation | GAC,GAG | D,E 1903 | NP_056056.2 | |
XM_005261262.3 | 6428 | Missense Mutation | GAC,GAG | D,E 1903 | XP_005261319.1 | |
XM_011546137.2 | 6428 | Missense Mutation | GAC,GAG | D,E 2110 | XP_011544439.1 | |
XM_011546139.2 | 6428 | Missense Mutation | GAC,GAG | D,E 2093 | XP_011544441.1 | |
XM_011546142.2 | 6428 | Missense Mutation | GAC,GAG | D,E 2076 | XP_011544444.1 | |
XM_011546146.2 | 6428 | Intron | XP_011544448.1 | |||
XM_017028865.1 | 6428 | Missense Mutation | GAC,GAG | D,E 2110 | XP_016884354.1 | |
XM_017028866.1 | 6428 | Missense Mutation | GAC,GAG | D,E 2110 | XP_016884355.1 | |
XM_017028867.1 | 6428 | Missense Mutation | GAC,GAG | D,E 2110 | XP_016884356.1 | |
XM_017028868.1 | 6428 | Missense Mutation | GAC,GAG | D,E 2110 | XP_016884357.1 | |
XM_017028869.1 | 6428 | Missense Mutation | GAC,GAG | D,E 2110 | XP_016884358.1 | |
XM_017028870.1 | 6428 | Missense Mutation | GAC,GAG | D,E 2110 | XP_016884359.1 | |
XM_017028871.1 | 6428 | Missense Mutation | GAC,GAG | D,E 2109 | XP_016884360.1 | |
XM_017028872.1 | 6428 | Missense Mutation | GAC,GAG | D,E 2093 | XP_016884361.1 | |
XM_017028873.1 | 6428 | Missense Mutation | GAC,GAG | D,E 2089 | XP_016884362.1 | |
XM_017028874.1 | 6428 | Missense Mutation | GAC,GAG | D,E 2082 | XP_016884363.1 | |
XM_017028875.1 | 6428 | Missense Mutation | GAC,GAG | D,E 2074 | XP_016884364.1 | |
XM_017028876.1 | 6428 | Missense Mutation | GAC,GAG | D,E 2064 | XP_016884365.1 | |
XM_017028877.1 | 6428 | Missense Mutation | GAC,GAG | D,E 2043 | XP_016884366.1 | |
XM_017028878.1 | 6428 | Missense Mutation | GAC,GAG | D,E 2005 | XP_016884367.1 | |
XM_017028879.1 | 6428 | Missense Mutation | GAC,GAG | D,E 1958 | XP_016884368.1 | |
XM_017028880.1 | 6428 | Missense Mutation | GAC,GAG | D,E 1958 | XP_016884369.1 | |
XM_017028881.1 | 6428 | Missense Mutation | GAC,GAG | D,E 1958 | XP_016884370.1 | |
XM_017028882.1 | 6428 | Missense Mutation | GAC,GAG | D,E 1937 | XP_016884371.1 | |
XM_017028883.1 | 6428 | Missense Mutation | GAC,GAG | D,E 1810 | XP_016884372.1 | |
XM_017028884.1 | 6428 | Missense Mutation | GAC,GAG | D,E 1658 | XP_016884373.1 | |
XM_017028885.1 | 6428 | Missense Mutation | GAC,GAG | D,E 1209 | XP_016884374.1 | |
XM_017028886.1 | 6428 | Missense Mutation | GAC,GAG | D,E 1209 | XP_016884375.1 | |
XM_017028887.1 | 6428 | Intron | XP_016884376.1 |