Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAACCATGTTTGCAGACTTGGATT[A/G]TGACATCGAAGAGGATAAACTGTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605926 | ||||||||||||||||||||
Literature Links: |
PICK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PICK1 - protein interacting with PRKCA 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039583.1 | 1 | Missense Mutation | TAT,TGT | Y,C 7 | NP_001034672.1 | |
NM_001039584.1 | 1 | Missense Mutation | TAT,TGT | Y,C 7 | NP_001034673.1 | |
NM_012407.3 | 1 | Missense Mutation | TAT,TGT | Y,C 7 | NP_036539.1 | |
XM_006724379.2 | 1 | UTR 5 | XP_006724442.1 | |||
XM_006724380.2 | 1 | UTR 5 | XP_006724443.1 | |||
XM_011530517.2 | 1 | Missense Mutation | TAT,TGT | Y,C 7 | XP_011528819.1 | |
XM_017029090.1 | 1 | UTR 5 | XP_016884579.1 | |||
XM_017029091.1 | 1 | UTR 5 | XP_016884580.1 | |||
XM_017029092.1 | 1 | UTR 5 | XP_016884581.1 | |||
XM_017029093.1 | 1 | UTR 5 | XP_016884582.1 | |||
XM_017029094.1 | 1 | UTR 5 | XP_016884583.1 | |||
XM_017029095.1 | 1 | UTR 5 | XP_016884584.1 |