Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGCGCGGGGACATGGGGCCCGAC[A/G]TGGAGCTGCCCAGCCACTCGAAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607712 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HIC2 - hypermethylated in cancer 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015094.2 | 200 | Missense Mutation | ATG,GTG | M,V 23 | NP_055909.2 | |
XM_011530007.2 | 200 | Missense Mutation | ATG,GTG | M,V 23 | XP_011528309.1 | |
XM_011530008.2 | 200 | Missense Mutation | ATG,GTG | M,V 23 | XP_011528310.1 | |
XM_011530009.2 | 200 | Missense Mutation | ATG,GTG | M,V 23 | XP_011528311.1 | |
XM_011530010.2 | 200 | Missense Mutation | ATG,GTG | M,V 23 | XP_011528312.1 | |
XM_017028669.1 | 200 | Missense Mutation | ATG,GTG | M,V 5 | XP_016884158.1 |